missing translation for 'onlineSavingsMsg'
Learn More

Bis(norbornadiene)rhodium(I) tetrafluoroborate

Product Code. p-7059131
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11450650

Brand: Thermo Scientific Alfa Aesar 044036.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36620-11-8 | C14H16BF4Rh | 373.99 g/mol

Bis(norbornadiene)rhodium(I) tetrafluoroborate is used in hydrogenation, Isomerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36620-11-8
Molecular Formula C14H16BF4Rh
Molecular Weight (g/mol) 373.99
MDL Number MFCD00671775
InChI Key HAYDJWBQWOEERB-UHFFFAOYSA-N
PubChem CID 10915722
SMILES [Rh+].F[B-](F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2

Specifications

Quantity 1 g
Sensitivity Air and moisture sensitive
Synonym bis norbornadiene rhodium i tetrafluoroborate, rh nbd 2bf4, bis eta-2,5-norbornadiene rhodium i tetrafluoroborate, bis norbornadiene tetrafluoroborato rhodium, bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate, rh nbd 2 bf4, bis norbomadiene rhodium i tetrafluoroborate, bis norbornadiene rhodium tetrafluoroborate
Solubility Information Fairly soluble in dichloromethane. Insoluble in water.
IUPAC Name λ¹-rhodium(1+) bis(bicyclo[2.2.1]hepta-2,5-diene) tetrafluoroboranuide
Formula Weight 373.99
Physical Form Powder
Chemical Name or Material Bis (norbornadiene)rhodium (I) tetrafluoroborate
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.