missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Bis(cyclopentadienyl)dimethylzirconium(IV), 98+%
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | ∽170°C (decomposition) |
Quantity | 1 g |
Formula Weight | 251.48 |
Percent Purity | ≥98% |
Chemical Name or Material | Bis(cyclopentadienyl)dimethylzirconium(IV) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'