Learn More
Bis-Biphenyl-4-yl-amine, TRC
Specifications
Specifications
| Chemical Name or Material | Bis-Biphenyl-4-yl-amine |
| CAS | 102113-98-4 |
| Molecular Formula | C24H19N |
| Quantity | 50 mg |
| Synonym | Bis(4-biphenyl)amine, [1,1'-Biphenyl]-4-amine, N-[1,1'-biphenyl]-4-yl-, Bis(4-biphenylyl)amine (6CI,7CI), N-[1,1'-Biphenyl]-4-yl[1,1'-biphenyl]-4-amine, Bis(1,1'-biphenyl-4-yl)amine, Bis(4-biphenyl)amine, Di([1,1'-biphenyl]-4-yl)amine, N,N-Bis(1,1'-biphenyl-4-yl)amine, N,N-Bis(4-biphenylyl)amine, N,N-Bis(4-phenylphenyl)amine |
| InChI Formula | InChI=1S/C24H19N/c1-3-7-19(8-4-1)21-11-15-23(16-12-21)25-24-17-13-22(14-18-24)20-9-5-2-6-10-20/h1-18,25H |
| SMILES | N(c1ccc(cc1)c2ccccc2)c3ccc(cc3)c4ccccc4 |
| IUPAC Name | 4-phenyl-N-(4-phenylphenyl)aniline |
| Molecular Weight (g/mol) | 321.41 |
| Formula Weight | 321.1517 |