Learn More
Bis(4-fluorophenyl) sulfone, 98+%
Product Code. 11381687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11381687 25 g 25g
11371687 5 g 5g
2 options
Product Code. 11381687
Brand: Thermo Scientific Alfa Aesar L04723.14
Request Bulk or Custom Format
CAS: 383-29-9 | C12H8F2O2S | 254.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 383-29-9
Molecular Formula C12H8F2O2S
Molecular Weight (g/mol) 254.25
MDL Number MFCD00000350
InChI Key PLVUIVUKKJTSDM-UHFFFAOYSA-N
Synonym 4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone
PubChem CID 67842
IUPAC Name 1-fluoro-4-(4-fluorophenyl)sulfonylbenzene
SMILES FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1
Specifications
Melting Point 97°C to 101°C
Odor Odorless
Quantity 25 g
Beilstein 1970911
Formula Weight 254.25
Percent Purity ≥98%
Chemical Name or Material Bis(4-fluorophenyl) sulfone