Learn More
Bis(2-ethylhexyl) Maleate, Mikromol™
Description
Bis(2-ethylhexyl) Maleate
Specifications
Specifications
| Chemical Name or Material | Bis(2 ethylhexyl)maleate |
| CAS | 142-16-5 |
| Analyte or Component Names | Bis(2 ethylhexyl)maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H36 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Bis(2-ethylhexyl)maleate |
| InChI Formula | InChI=1S/C20H36O4/c1-5-9-11-17(7-3)15-23-19(21)13-14-20(22)24-16-18(8-4)12-10-6-2/h13-14,17-18H,5-12,15-16H2,1-4H3/b14-13- |
| SMILES | CCCCC(CC)COC(=O)\C=C/C(=O)OCC(CC)CCCC |
| IUPAC Name | bis(2-ethylhexyl) (Z)-but-2-enedioate |
| Show More |