Learn More
bis(2-Chloroethyl) ether 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30311383
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311383 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30311383 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011038ME
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis-(2-chloroethyl)ether
CAS 111-44-4
Analyte or Component Names Bis-(2-chloroethyl)ether
Shipping Condition Room Temperature
Molecular Formula C4 H8 Cl2 O
Concentration 1000 μg/ml
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C4H8Cl2O/c5-1-3-7-4-2-6/h1-4H2
SMILES ClCCOCCCl
IUPAC Name 1-chloro-2-(2-chloroethoxy)ethane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 143.01
Quantity 1 mL
Formula Weight 141.9952 g/mol
Mehr anzeigen Weniger anzeigen