missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals

Catalog Number p-3212418
Click to view available options
Quantity:
1 g
5 g
25 g
1295-35-8
C16H24Ni
275.06
MFCD00058902
JRTIUDXYIUKIIE-KZUMESAESA-N
bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018
6433264
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
1295-35-8
C16H24Ni
275.06
MFCD00058902
JRTIUDXYIUKIIE-KZUMESAESA-N
bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018
6433264
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 60.0°C
Color Golden
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;nickel
Formula Weight 275.06
Physical Form Shiny Crystalline Powder
Chemical Name or Material Bis(1, 5-cyclooctadiene)nickel(0)
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bis(1,5-cyclooctadiene)nickel(0)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H228-Flammable solid.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'