missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-sulfonyl chloride, 97%

Product Code. 11480117
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11480117 25 g 25g
11470117 5 g 5g
11490117 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11480117

Brand: Thermo Scientific Alfa Aesar B21031.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1623-93-4 | C12H9ClO2S | 252.71 g/mol

It is a reactant for preparation of anilinopyrimidinesulfonamides as potential anticancer agents, synthesis of inhibitors of the hypoxia inducible factor pathway. It participates in the preparation of hydroxy-sulfonyl-piperidinyl butyramides as HDAC inhibitors and potential antitumors and is also involved in palladium-catalyzed desulfitative C-arylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for preparation of anilinopyrimidinesulfonamides as potential anticancer agents, synthesis of inhibitors of the hypoxia inducible factor pathway. It participates in the preparation of hydroxy-sulfonyl-piperidinyl butyramides as HDAC inhibitors and potential antitumors and is also involved in palladium-catalyzed desulfitative C-arylation.

Solubility
Reacts with water.

Notes
Store at room temperature. Keep away from moisture, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1623-93-4
Molecular Formula C12H9ClO2S
Molecular Weight (g/mol) 252.71
MDL Number MFCD00078658
InChI Key ALBQXDHCMLLQMB-UHFFFAOYSA-N
Synonym 4-biphenylsulfonyl chloride, 1,1'-biphenyl-4-sulfonyl chloride, biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulphonyl chloride, 4-biphenylsulfonylchloride, 1,1'-biphenyl-4-sulfonylchloride, biphenyl-4-sulphonyl chloride, 4-phenylbenzene-1-sulfonyl chloride, 1,1-biphenyl-4-sulfonylchloride, pubchem20990
PubChem CID 74192
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 112°C to 117°C
Boiling Point 200°C to 205°C (6 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 2694373
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name 4-phenylbenzenesulfonyl chloride
Formula Weight 252.72
Percent Purity 97%
Chemical Name or Material Biphenyl-4-sulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.