Learn More
Biperiden Hydrochloride, Mikromol™
Description
Biperiden Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Biperiden hydrochloride |
| CAS | 1235-82-1 |
| Analyte or Component Names | Biperiden hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H29 N O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Biperiden hydrochloride, (1RS)-1-[(1RS,2SR,4RS)-Bicyclo[2.2.1]hept-5-en-2-yl]-1-phenyl-3-(piperidin-1-yl)propan-1-ol hydrochloride, Akineton hydrochloride, Akinophyl, Biperidene hydrochloride, NSC 170950, NSC 84989, Tasmolin, 1-Piperidinepropanol, alpha-bicyclo[2.2.1]hept-5-en-2-yl-alpha-phenyl-, hydrochloride (1:1), 1-Piperidinepropanol, alpha-5-norbornen-2-yl-alpha-phenyl-, hydrochloride (6CI,7CI,8CI), 1-Piperidinepropanol, alpha-bicyclo[2.2.1]hept-5-en-2-yl-alpha-phenyl-, hydrochloride (9CI), alpha-(Bicyclo[2.2.1]hept-5-en-2-yl)-alpha-phenyl-1-piperidinepropanol hydrochloride |
| InChI Formula | InChI=1S/C21H29NO.ClH/c23-21(19-7-3-1-4-8-19,11-14-22-12-5-2-6-13-22)20-16-17-9-10-18(20)15-17;/h1,3-4,7-10,17-18,20,23H,2,5-6,11-16H2;1H/t17-,18+,20+,21+;/m1./s1 |
| SMILES | Cl.O[C@](CCN1CCCCC1)([C@H]2C[C@H]3C[C@@H]2C=C3)c4ccccc4 |
| IUPAC Name | (1R)-1-[(1R,4R,5S)-5-bicyclo[2.2.1]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride |
| Show More |