Learn More
Biperiden hydrochloride, MedChemExpress
Product Code. 16476106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476106 100 mg 100mg
16441656 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16476106 Supplier Medchem Express Supplier No. HY13204
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Biperiden (KL 373) hydrochloride is a non-selective muscarinic receptor antagonist that competitively binds to M1 muscarinic receptors, thereby inhibiting acetylcholine and enhancing dopamine signaling in the central nervous system. Biperiden hydrochloride has the potential for the research of Parkinson's disease and other related psychiatric disorders.
Biperiden (KL 373) hydrochloride is a non-selective muscarinic receptor antagonist that competitively binds to M1 muscarinic receptors, thereby inhibiting acetylcholine and enhancing dopamine signaling in the central nervous system. Biperiden hydrochloride has the potential for the research of Parkinson's disease and other related psychiatric disorders.
Specifications
Chemical Name or Material Biperiden hydrochloride
Physical Form Solid
CAS 1235-82-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H30ClNO
For Use With (Application) Neuroscience-Neurodegeneration
Synonym KL 373 hydrochloride
Solubility Information DMSO : 20 mg/mL (57.48 mM; Need ultrasonic) ∣H2O : 5 mg/mL (14.37 mM; Need ultrasonic)
SMILES OC(C1CC2C=CC1C2)(CCN3CCCCC3)C4=CC=CC=C4.[H]Cl
Molecular Weight (g/mol) 347.92
Formula Weight 347.92
Grade Research