Learn More
Lonza™ BioWhittaker™ General-Purpose Serum-Free Media
Chemically defined media optimized for specific cell types
Brand: Lonza™ 77232
Additional Details : Weight : 1.00000kg
Description
PC-1™
- Provides complete media system, including PC-1 Supplement for cultivation of primary cells and anchorage-dependent cell lines
- Contains complete HEPES buffering system
- Designed for growth of adherent and nonadherent cells
Specifications
2° to 8°C | |
Serum Free Medium (SFM) | |
No Glutamine | |
2 x 500 mL |