Learn More
ACROBiosystems Biotinylated Mouse CD3 epsilon&CD3 gamma Heterodimer Protein, His, Avitag&Flag Tag (MALS verified)
Brand: ACROBiosystems CDG-M58D2-25ug
Additional Details : Weight : 0.00010kg
Description
T-cell surface glycoprotein CD3 delta & CD3 gamma chain, also known as CD3D & CD3G or CD3D&CD3G respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Biotinylated Mouse CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.8 kDa (CD3E) and 15.2 kDa (CD3G). Subunit CD3E carries a polyhistidine tag at the C-terminus, followed by an AviSpecifications
NP_031674.1/NP_033980.1 | |
25 μg | |
Asp 23 - Asp 108 (CD3E) & Gln 23 - Ser 116 (CD3G) | |
CD3 epsilon & CD3 gamma,CD3E & CD3G | |
Precision Avi | |
His Tag & Avi Tag & Flag Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE. |
16.8 kDa (CD3E) & 15.2 kDa (CD3G) | |
Biotinylated Mouse CD3E&CD3G; Heterodimer Protein, His,AviTag&Flag; Tag (CDG-M58D2) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 108 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # P22646-1 (CD3E) & P11942-1 (CD3G)). | |
-20°C | |
CD3E & CD3G | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |