Learn More
ACROBiosystems Biotinylated Human ROR2 / NTRKR2 Protein, His, Avitag™
Biotinylated Protein
Brand:  ACROBiosystems RO2-H82E3-200ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16448019
  • 19250.81 NOK / 200µg
Estimated Shipment: 17-01-2023
to see stock.
Description
Description
Tyrosine-protein kinase transmembrane receptor ROR2 is also known as Neurotrophic tyrosine kinase, receptor-related 2 (NTRKR2), which belongs to the protein kinase superfamily and Tyr protein kinase family and ROR subfamily. ROR2 is a homodimer protein, which can binds YWHAB or interact with WTIP. ROR2 may be involved in the early formation of the chondrocytes. It seems to be required for cartilage and growth plate development.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
AAI30523.1
45.0 kDa
200 µg
Glu 34 - Gly 403
ROR2,NTRKR2
Precision Avi
PBS, pH7.4
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Biotinylated Human ROR2, His,Avitag (RO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # A1L4F5-1).
-20°C
ROR2
His Tag and Avi Tag
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins