Learn More
IBA Lifesciences Biotin
Product Code. 12863206 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
2 g
5 g
Unit Size:
2g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12863206 5 g 5g
12853206 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12863206 Supplier IBA Lifesciences Supplier No. 21016005
  • ONLINE EXCLUSIVE PRICE
    2340.00 NOK
    2105.47 NOK / 5g
    Save 234.53 NOK
    10% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Biotin powder for preparation of Strep-Tactin™XT elution buffer
  • Biotin is a water-soluble B-complex vitamin, also known as vitamin B7 or vitamin H. It binds to the biotin-binding pockets of streptavidin, Strep-Tactin™, and Strep-Tactin™XT with different affinities.
  • It allows the efficient elution of Strep-tag™II or Twin-Strep-tag™ fusion proteins from Strep-Tactin™XT, while maintaining the reusability of the resin.
  • It can be used for preparation of Strep-Tactin™XT elution buffer, Buffer BXT.
  • In addition, biotin can be used for elution of biotinylated proteins from Strep-Tactin™, where it binds irreversibly to the resin - Strep-Tactin™ cannot be regenerated in this kind of application.
Specifications
Quantity 5 g
For Research Use Only.