Learn More
Agilent BioTek Washer Biomagnetic Separation Carrier Accessories
GREENER_CHOICE
Product Code. 11997411 Shop All Agilent BioTek Products
Click to view available options
Type:
96-Well Ring Magnet
Unit Size:
Each
Product Code. 11997411
Brand: Agilent BioTek 7102216
Enables high strength biomagnetic separation
  • Magnet ring
  • External
  • For insertion into washer biomagnetic separation carriers
  • Beads pulled to 4-zone ring at well bottom
TRUSTED_SUSTAINABILITY
Specifications
Type 96-Well Ring Magnet
For Use With (Equipment) 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer