Learn More
Agilent BioTek Liquid Handling Control Software
GREENER_CHOICE
Product Code. 15396196 Shop All Agilent BioTek Products
Click to view available options
:
LHC Software
Product Code. 15396196
Brand: Agilent BioTek LHC2SECURE
Includes
Software on CD, installation guide and 5 licenses
Full PC control provided by LHC Software leads to increased convenience when programming important assay-specific protocol requirements in a familiar Windows environment. Fisher Scientific - Liquid Handling Control Software is ideal for use with 405TM Touch Washer, 405TM LS Washer, ELx405TM Select Deep Well Washer, EL406TM Washer Dispenser and MultiFloTMFX Dispenser.
  • Intuitive StepWise protocol creation with an unlimited number of methods included for ultimate flexibility
  • Automatic protocol validation for error-free assurance
  • Dual mode control to create and run protocols from PC or download to instrument keypad
  • Security enabled protection for downloaded protocols with LHC2 SECURE
  • Printed protocol reports and automatic protocol history
  • Custom maintenance reminders for continued instrument performance in busy laboratories
  • Multiple instruments controlled in parallel, including BioStack integrations for unattended batch processing
Compliance
United States Food and Drug Administration 21 CFR Part 11 regulation (LHC Secure Edition)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 21 CFR Part 11
Includes Software on CD, installation guide and 5 licenses
For Use With (Equipment) BioTek 405 Touch Washer, 405 LS Washer, ELx405 Select Deep Well Washer, EL406 Washer Dispenser and MultiFlo Dispenser
Type Liquid Handling Control Software
Product Content Correction
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt