Learn More
Sartorius™ Biosart™ Monitor
Product Code. 10679792 Shop All Sartorius Products
Change view
Click to view available options
Color:
Gray/White
Green/Dark Green
White/Black
Packaging:
2 layers of 24
4 bags of 12
48/Pk.
Unit Size:
Pack of 48
Product Code. Color Packaging unitSize
10679792 White/Black 48/Pk. Pack of 48
11823292 Green/Dark Green 4 bags of 12 Pack of 48
11832691 Gray/White 4 bags of 12 Pack of 48
11833292 Gray/White 2 layers of 24 Pack of 48
4 options
Product Code. 10679792
Brand: Sartorius™ 164034706ACK
Microbiological and analytical filtration units offering superior performance
The system enables the user to select the optimal hardware and consumables for their needs in quality assurance. At the heart of the system is a stainless steel manifold designed to accommodate all types of filter holder and funnels. Please note: grey filters turn black when wet. Multi-branch system for routine colony or particle counting.
TRUSTED_SUSTAINABILITY
Specifications
Color White/Black
Sterility Sterile
No. per Case 48Pack
No. per Pack 48Pack
Packaging 48/Pk.
Quantity 48Pack
Type Monitor
compliance-icons
Product Identifier
  • X48 BIOSART100 CN G/W ,45µ IND
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose