Learn More
Thermo Scientific™ Bioreactor Probe Assembly for use with Single-Use Bioreactors
Shop All Thermo Scientific ProductsDescription
- The Thermo Scientific™ Bioreactor Probe Assembly (nonsterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ Single-Use Bioreactors.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Silicone |
| For Use With | HyPerforma™ Single-use Bioreactors, DynaDrive™ Single-use Bioreactors |
| Includes | Aseptiquick G Connector |
| Color | Clear |
| Product Type | Probe Assembly |