Learn More
3M™ Biomass Detection Cuvette BTCUV
Product Code. 17203744
Change view
Click to view available options
Test Type: Environmental Test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17203744 Test Type: Environmental Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17203744 Supplier 3M™ Supplier No. 7000138443
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cuvettes for aqueous solution samples for use with the 3M™ Clean-Trace™ Biomass Detection Kit.
  • Gives real-time indication of total biological load allowing immediate corrective action.
  • Offers flexibility when dealing with challenging samples.
  • Ideal for process water, wastewater, water tanks and cooling towers.
Specifications
Product Type Accessory
Test Type Environmental Test
For Use With (Equipment) 3 M™ Clean-Trace™ Biomass Detection Kit
Quantity 1000/Cs.