Learn More
Sartorius™ Biohit™ Proline Multi-Channel Mechanical Pipettes
Ideal for universities and colleges or any laboratory seeking cost-efficient, reliable liquid-handling tool. Sartorius™ Biohit™ Proline Multi-Channel Mechanical Pipettes are adjustable-range pipettes that can be used with many tip brands. Available in 8- or 12-channel designs.
Brand: Sartorius™ 720310
9532.91 NOK valid until 2024-12-31
Use promo code "23285" to get your promotional price.
Additional Details : Weight : 0.17500kg
Description
- Light weight with high accuracy and precision for use by professionals
- Wide range of pipettes with adjustable volume ranges
- Easy volume setting with click stop mechanism
- Relaxed grip with ergonomic finger support
- Reduced risk of contamination with Safe-Cone Filters available
- Compatible with many universal tips, including Optifit Tips and SafetySpace Tips
Specifications
CE | |
Gray | |
Relaxed | |
12 | |
Proline Mechanical Pipette | |
For universities and colleges, or any laboratory seeking a cost-efficient liquid handling tool. | |
0.5 to 10μL | |
1 Each |
For 10μL Optifit Tips And 10μL SafetySpace™ Filter Tips | |
10 μL Optifit™ Tip and 10 μL SafetySpace™ Filter Tip | |
Polypropylene | |
Biohit™ Proline™ | |
0.5 to 10 μL | |
For 10μL Optifit Tips And 10μL SafetySpace™ Filter Tips | |
0.1μL |