Learn More
Sartorius™ Biohit™ Proline™ Fixed Volume Pipettes
Product Code. p-8382012 Shop All Sartorius Products
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
2000 μL
5000 μL
Unit Size:
Each
Product Code. 10063481
Brand: Sartorius™ 722004
Only null left
Add to basket
Add to basket
Single channel fixed volume pipettes.
Single channel fixed volume pipettes.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy 0.8μL
Certifications/Compliance ISO 3696
ClipTip Optifit Tips, Non-filtered, Filter Tips
Compatible Tips Optifit Tips, Non-filtered, Filter Tips
Autoclavable Autoclavable
For Use With Laboratory
Number of Channels 1
Sterility Sterile
Type Fixed Volume Pipette
Volume (Metric) 10 μL
Filter Type Safe-Cone Filters
Finger Style Ergonomic
For Use With (Application) It is particularly designed for repetitive and long-lasting pipetting in order to prevent work related hand, arm and shoulder disorders or Repetitive Strain Injury (RSI).
For Use With (Equipment) Laboratories
Precision 0.8%
Quantity 1 Each
Repeatability < 0,20
Resistance Chemical
Warranty 15 Months
Weight (Metric) 77 g