Learn More
Macherey-Nagel™ BioFix™ Lumi-10 Compact Luminometer
Product Code. 10716722 Shop All Macherey-Nagel Products
Change view
Click to view available options
Range:
380 nm to 630 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10716722 380 nm to 630 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10716722 Supplier Macherey-Nagel™ Supplier No. 940008
  • ONLINE EXCLUSIVE PRICE
    127030.00 NOK
    117769.12 NOK / Pack of 1
    Save 9260.88 NOK
    7% Off
Estimated Shipment: 18-11-2026
For the measurement of bio and chemical luminescence reactions with constant light emission.
  • Compact luminometer for mobile use
    • Ideally suited for use in the laboratory or on the road and can be operated with a power supply as well as rechargeable batteries
  • Incredibly versatile
    • Thanks to its highly sensitive detector (Ultra‐Fast Single Photon Counter) the BioFix™ Lumi‐10 can be used for a variety of applications including, but not limited to, bio toxicity tests, ATP‐ and biomass determinations, reporter‐gene assays, luminescence immunoassays, and NAD(P)H measurements
  • Individually programmable
    • The BioFix™ Lumi‐10 has six individually adjustable measurement protocols and a data memory for up to 2000 results
    • Provides the opportunity for single, multiple and extensive screening measurements
    • Results are optionally displayed in % inhibition, % stimulation or RLU (relative light units)
    • User can set particular measurement parameters such as incubation time or measurement time individually
    • By a previous definition of detection limits, the results can be automatically classified by the device
    • Pre‐programmed test methods are available for the determination of luminescent bacteria toxicity tests and ATP tests
  • Includes manual, rack, cuvettes, and spare adapter
Specifications
Display Type Backlit graphic display (128 x 64 pixels)
Connection Type RS232 interface for data transfer to the PC or printer
Range 380 nm to 630 nm
Voltage 115/230 V
Frequency 50/60 Hz
Weight (Metric) 2 kg (with batteries)
Includes Manual, rack, cuvettes, and spare adapter, 3 Rechargeable batteries: NiCd R14/C /Baby/UM2 batteries; 1600 mAh, 3 preprogrammed tests, 6 free programmable methods, % inhibition, %stimulation, RLU
Certifications/Compliance CE certified