Learn More
Cytiva Biodyne™ Plus Membrane
Product Code. 17174511
Change view
Click to view available options
Dimensions (LxW):
132 x 132 mm
20 cm x 20 m
30 cm x 3 m
Unit Size:
1 roll
10 rolls
Pack of 50
Product Code. Dimensions (LxW) unitSize
17174511 132 x 132 mm Pack of 50
17184511 30 cm x 3 m 1 roll
17164511 20 cm x 20 m 10 rolls
3 options
Product Code. 17174511
Brand: Cytiva 60404
Only null left
Add to basket
Add to basket
Biodyne Plus membrane, like Biodyne B membrane, contains pore surfaces populated by a high density of quaternary ammonium groups.
  • Biodyne Plus membrane is recommended for detection systems that produce high background on Biodyne B membrane
TRUSTED_SUSTAINABILITY
Specifications
Packaging Type Disc
Description Non-sterile
Material Positively-charged Nylon 6.6
Pore Size 0.45 μm
Dimensions (LxW) 132 x 132 mm
Thickness 5.7 to 6.7 μm