Learn More
Cytiva Biodyne™ C Membrane
Product Code. 17124511
Change view
Click to view available options
Dimensions (LxW):
20 cm x 3 m
20 x 20 cm
7 x 8.5 cm
Unit Size:
1 roll
Pack of 10
Product Code. Dimensions (LxW) unitSize
17124511 20 x 20 cm Pack of 10
17134511 7 x 8.5 cm Pack of 10
17154511 20 cm x 3 m 1 roll
3 options
Product Code. 17124511
Brand: Cytiva 60314
Only null left
Add to basket
Add to basket
Biodyne C membrane is negatively charged with pore surfaces populated by a high density of carboxyl groups.
  • The membrane can be derivatized by coupling reactions with these groups
  • It is ideal for reverse dot blots and affinity purification
TRUSTED_SUSTAINABILITY
Specifications
Packaging Type Sheets
Description Non-sterile
Material Negatively-charged Nylon 6.6
Pore Size 0.45 μm
Dimensions (LxW) 20 x 20 cm
Thickness 11 to 13 μm