Learn More
Corning™ BioCoat™ Endothelial Cell Growth Environment Kit
GREENER_CHOICE
Product Code. 11523590
Change view
Click to view available options
Type:
Complete Endothelial Cell Growth Environment Kit
Endothelial Cell Culture Medium
Unit Size:
1 set
Product Code. Type unitSize
11523590 Endothelial Cell Culture Medium 1 set
10365771 Complete Endothelial Cell Growth Environment Kit 1 set
2 options
Product Code. 11523590
Brand: Corning™ 355054
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Complete Environment Kit: 500mL BD Endothelial Cell Culture Medium (MCDB 131 medium containing hydrocortisone, heparin and 2% fetal bovine serum; 5μg lyophilized Mouse Epidermal Growth Factor (EGF), receptor grade; 100mg lyophilized Bovine Endothelial Cell Growth Supplement (ECGS); 50mg lyophilized Soybean Trypsin Inhibitor; Five BD BioCoat Type I Collagen 75cm2 Flasks
Increases endothelial cell yield from a variety of sources
Ideal for human umbilical vein (HUVEC), pulmonary artery (HPAEC), and aorta (HAEC), as well as fetal bovine heart (FBHEC) cells. Interactions between components of the extracellular matrix and growth factors play a crucial role in determining the adaptation and responses of mammalian cells to their micro environments both in vivo and in vitro.
  • Cells can be subcultured according to routine procedures, seeded into fermentation systems, or used in a variety of bioassays
  • Integrate specially formulated low-serum or serum-free medium, culture supplements and growth factors
  • Each environment lot is tested in a bioassay and all components are tested for bacteria and fungi
These products are for research use only and are not intended for diagnostic purposes.
Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology
Order Info
Media and cellware are also available separately.
TRUSTED_SUSTAINABILITY
Specifications
Type Endothelial Cell Culture Medium
For Use With (Application) Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology
Storage Requirements 2° to 8°C
No. per Pack 1/Pk.
No. per Case 1/Ea.