Learn More
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Product Code. 10479392
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
Unit Size:
Pack of 20
Product Code. Diameter (Metric) unitSize
10479392 35 mm Pack of 20
10142861 60 mm Pack of 20
2 options
Product Code. 10479392
Brand: Corning™ 354459
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
  • Source: Mouse
  • TC-treated
  • Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Round
Material Polystyrene
Shape Round
Surface Treatment Collagen Type IV
Type Cell Culture Dish
Diameter (Metric) 35 mm