Learn More
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
GREENER_CHOICE
Product Code. 10142861
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
Unit Size:
Pack of 20
Product Code. Diameter (Metric) unitSize
10142861 60 mm Pack of 20
10479392 35 mm Pack of 20
2 options
Product Code. 10142861
Brand: Corning™ 354416
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
  • Source: Mouse
  • TC-treated
  • Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Round
Culture Area 21.3 cm2
Material Polystyrene
Shape Round
Surface Treatment Collagen Type IV
Type Cell Culture Dish
No. per Case 20/Cs.
No. per Pack 5/Pk.
Diameter (Metric) 60 mm