Learn More
Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask
Click to view available options
Capacity (Metric):
70 mL
250 mL
750 mL
Culture Area:
175 cm2
25 cm2
75 cm2
Unit Size:
Pack of 10
Pack of 5
Description
- BioCoat™ provides a superior solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Specifications
Specifications
| Closure Material | Phenolic |
| Capacity (Metric) | 750 mL |
| For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
| Neck Style | Canted |
| Shape | Rectangular |
| Surface Coating | Collagen IV |
| Sterility | Sterile |
| Closure Type | Plug Seal |
| Culture Area | 175 cm2 |