Learn More
Nitritex™ BioClean Permaflow™ Sterile Cleanroom Marker Pens
Product Code. 15994841
Change view
Click to view available options
Color:
Black
Blue
Red
Description:
Bagged
Double bagged
Double bagged, Pack of 10
Unit Size:
Pack of 10
Pack of 200
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Description Unit Size
15994841 Black Double bagged Pack of 10
15298308 Black Bagged Pack of 200
15365905 Blue Double bagged, Pack of 10 Pack of 10
15278308 Blue Double bagged Pack of 200
15218318 Red Bagged Pack of 200
15904851 Red Double bagged Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15994841 Supplier Nitritex™ Supplier No. SBPFPBK1
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 23-11-2026
Processed to ensure ISO Class 4 & EU GMP Grade A compatibility. Nitritex™ BioClean Permaflow™ Cleanroom Marker Pens are individually packaged and available with black, blue, or red ink. Double-bagged varieties are available.
  • Super permanent ink in black, blue, or red
  • Fade and water resistant
  • Quick drying
  • Non-toxic
  • Alcohol resistant labeling
  • Autoseal ink prevents drying
  • Store in a dry, cool place away from direct sunlight and fluorescent lighting
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Processed in an NEBB certified ISO Class 4 cleanroom environment
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Description Double bagged
Length (English) 5.5 in.
Length (Metric) 140 mm