Learn More
Ansell BIOCLEAN™ Permaflow Irradiated Cleanroom Ballpoint Pen S-BPBP-1
Product Code. 30040334
Change view
Click to view available options
Color:
Black
Blue
Unit Size:
Pack of 300
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30040334 Black Pack of 300
30040335 Blue Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30040334 Supplier Ansell Supplier No. SBPBP1BK
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Ballpoint pens for alcohol resistant labelling are processed to ensure ISO Class 4 and EU GMP Grade A compatibility, fade and water resistant, quick drying, and non-toxic.
  • BioClean Permaflow cleanroom pens have fade and water resistant fast drying permanent ink, and feature an alcohol resistant label for easy cleaning within a critical environment, they are ISO Class 4 and EU GMP Grade A compatible.
  • With a robust, lightweight, tinted polypropylene round barrel and 1 mm point size for quality writing precision. Ink complies with documentary use standard ISO 12757-2
TRUSTED_SUSTAINABILITY
Specifications
Tip Style Rollerball
Product Type Ballpoint Pen
Color Black