Learn More
Ansell BioClean™ BioClean-C Chemotherapy Sterile Protective Apron
Product Code. p-7162371
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 50
Product Code. 15218278
Brand: Ansell SBCDAS
Only null left
Add to basket
Add to basket
Sterile aprons provide protection against a range of chemotherapy drugs.
  • Tested against permeation standard ASTM F739-12
  • Tie tapes at rear
  • Adjustable neck fastening
  • Lightweight low-linting CleanTough material
  • Silicone-free
  • Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications
  • Conforms to Category 3 - Personal Protective Equipment Regulation (EU) 2016/425
  • Tested against standard EN ISO 13034:2005 + A1:2009 protective clothing against liquid chemicals (Type PB [6])
  • Tested against standard EN 14325 protective clothing against chemicals
  • Tested against permeation standard ASTM F739-12
  • Processed in an NEBB certified ISO Class 4 environment
  • Tested against standards: EN 530, Abrasion resistance; EN 6530, Resistance to penetration by liquids; EN 13934-1, Tensile strength; EN 9073-4, Trapezoidal tear resistance; EN 863, Puncture resistance; and EN 13935-2, Seam strength
  • Sterilized in accordance with ISO 11137-1:2006
  • Test: Abrasion Resistance - Result: 10 to 100 cycles - Performance class (EN 14325):1
  • Test: Trapezoidal tear Resistance - Result: MD 50.6 N - Performance class (EN 14325):3
  • Test: Trapezoidal tear Resistance - Result: CD 32.3 N - Performance class (EN14325):2
  • Test: Puncture Resistance - Result:10 N - Performance class (EN14325):1
TRUSTED_SUSTAINABILITY
Specifications
Type Chemotherapy Protective Apron
Size Small
Color Blue
Disposable Single-use
Closure Type Waist Ties
Sterility Sterile
Latex Free No
Neck Style Adjustable
For Use With (Application) Critical and Controlled Environments
Three (3) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.