Learn More
Novacyt - Microgen Bioproducts Biochemical Identification System: Bacillus spp.
Product Code. 15938022 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
20 Tests
Unit Size:
20 tests
Product Code. Quantity unitSize
15938022 20 Tests 20 tests
1 options
Product Code. 15938022
Brand: Novacyt - Microgen Bioproducts MID66CE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Biochemical identification system for Bacillus spp. from foods and other environmental samples.
  • Twenty-four-substrate biochemical identification system.
  • From a range of different technologies for the specific detection of Bacillus spp. from foods and other environmental samples.
  • From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
  • These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
  • The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
For Use With (Application) Identification of Bacillus Spp.
Quantity 20 Tests