Learn More
Thermo Scientific™ BioBasic™ 18 HPLC Columns
Product Code. 16318616 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4 mm
4.6 mm
20 mm
Unit Size:
Each
Pack of 4
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
16318616 250 mm 20 mm Each
10434773 250 mm 4.6 mm Each
10151213 250 mm 2.1 mm Each
10097534 150 mm 1 mm Each
10435553 50 mm 1 mm Each
10181113 150 mm 2.1 mm Each
10293113 100 mm 2.1 mm Each
10475173 50 mm 2.1 mm Each
10502645 150 mm 4.6 mm Each
10735706 250 mm 1 mm Each
10160703 100 mm 1 mm Each
11558310 10 mm 4 mm Pack of 4
11548310 10 mm 2.1 mm Pack of 4
10448674 10 mm 1 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 16318616 Supplier Thermo Scientific™ Supplier No. 72105259270A
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Use this 300Å wide-pore silica for the separation of small to medium proteins and other biomolecules.
Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.
  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations
TRUSTED_SUSTAINABILITY
Specifications
Column Format Preparative Column, Standard
Diameter (Metric) 20 mm
Length (Metric) 250 mm
Max. Pressure 4300 psi (295 bar)
Particle Size 5 μm
pH 2 to 9
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase