Learn More
Macherey-Nagel™ BIO-LAB-TOP
Product Code. p-8013718
Change view
Click to view available options
Dimensions (L x W):
48 x 60 cm
5000 x 48 cm
5000 x 60 cm
60 x 1000 cm
No. per Pack:
1 Ea.
100Pack
Unit Size:
Each
Pack of 100
Product Code. Dimensions (L x W) No. per Pack unitSize
10789531 48 x 60 cm 100Pack Pack of 100
10344461 5000 x 48 cm 1 Ea. Each
10542323 60 x 1000 cm 1 Ea. Each
10420053 5000 x 60 cm 1 Ea. Each
4 options
Product Code. 10789531
Brand: Macherey-Nagel™ 740801
Protection of laboratory surfaces from spills.
  • Filter paper, one side coated with polyethylene
Specifications
Product Type Surface Protection Paper
Dimensions (L x W) 48 x 60 cm
Thickness 0.22 mm
No. per Pack 100Pack
Quantity 100Pack