Learn More
Macherey-Nagel™ BIO-LAB-TOP
Product Code. 10542323
Click to view available options
Dimensions (L x W):
48 x 60 cm
5000 x 48 cm
5000 x 60 cm
60 x 1000 cm
No. per Pack:
1 Ea.
100Pack
Unit Size:
Each
Pack of 100
Product Code. 10542323
Brand: Macherey-Nagel™ 740821
Protection of laboratory surfaces from spills.
  • Filter paper, one side coated with polyethylene
Specifications
Product Type Surface Protection Paper
Dimensions (L x W) 60 x 1000 cm
Thickness 0.22 mm
No. per Pack 1 Ea.
Quantity 1 Ea.