Learn More
Nitritex™ Bio-Clean™ D Disposable Coverall with Hood
BIOPHARMA
Product Code. p-7083195
Change view
Click to view available options
Size:
3X-Large
4X-Large
Size (Metric) Chest:
124 to 132 cm
132 to 140 cm
Height (Metric):
194 to 200 cm
200 to 206 cm
Unit Size:
Pack of 15
Product Code. Size Size (Metric) Chest Height (Metric) unitSize
15268128 3X-Large 124 to 132 cm 194 to 200 cm Pack of 15
15278128 4X-Large 132 to 140 cm 200 to 206 cm Pack of 15
2 options
Product Code. 15268128
Brand: Nitritex™ BDCHT3XL
Only null left
Add to basket
Add to basket
Includes
Front zip, with protective flap, elasticated hood
Provides exceptional comfort, protection, even when handling cytotoxic drugs.
  • Low linting
  • Thumb loop to ensure a secure hold
  • Elasticated openings to restrict particle release
  • Prevent bellows effect due to air pressure build-up
  • Bound seams, high quality zip with adhesive material flap
  • Overboots have slip-resistant soles and reinforced ties
  • Antistatic
Certifications
ISO Class 4
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material CleanTough™ Fabric
Color White
Disposable Single-use
Cuff Style Elastic
Hood Type Elastic
Seam Type Bound
Sterility Non-sterile
Back Style Elastic
Certifications/Compliance ISO 9001:2015 and (EU)2016/425, EN ISO 13982-1:2004+A1:2010, EN ISO 13034:2005+A1:2009, EN ISO 14325, ASTM F739-12, EN 1149-5:2008, EN530, EN 7854, EN 6530, EN 13934-1, EN9073-4, EN 863 and EN 13935-2
Description Bound seams with single needle stitching
Size (English) Chest 48 to 51 in.
Size (Metric) Chest 124 to 132 cm
For Use With (Application) Controlled Environments
Height (English) 76 to 78 in.
Height (Metric) 194 to 200 cm
Quantity 15/Cs.
Packaging Individually Packaged