Learn More
Micronova BinLiners™
Product Code. p-200115225 Shop All Micronova Products
Change view
Click to view available options
Dimensions (L x W x H):
6 x 46 x 36 in.
40 x 20 x 14 in.
48 x 40 x 48 in.
Unit Size:
Pack of 24
Pack of 35
Pack of 50
Product Code. Dimensions (L x W x H) unitSize
30533190 6 x 46 x 36 in. Pack of 24
30533189 40 x 20 x 14 in. Pack of 50
30533191 48 x 40 x 48 in. Pack of 35
3 options
Product Code. 30533190
Brand: Micronova BL38265BBMW1032
Designed for cleanroom material disposal and transfer with low density polyethylene liners
  • Micronova BinLiner Bags are made from FDA compliant, low density polyethylene and are designed for the collection and disposal of waste, garment collection, storage, protective covers for equipment and the collection and transport of parts or raw materials in cleanrooms and controlled environments
  • Low density polyethylene liners in a range of sizes
  • Thicknesses range from 1 mL to 6 mL, and sizes from 1 gal capacity to 60 gal
  • The 40 x 48 in. and 40 x 60-inch liners work with the BLSSH-1624 BinLiner Stand
  • The 34 x 40 in. liners work with the BLSSH-1515 BinLiner Stand
  • Liners are produced for use in clean and controlled environments
  • Liner colors (tints) include transparent, blue, and gray
Specifications
Content And Storage 15°C to 25°C
Autoclavable Not Autoclavable
Capacity (English) 400 gal.
Capacity (Metric) 1514 L
Certifications/Compliance ISO 9001 Certified Facility
Closure Type None
Color Clear
Disposable Single-use
For Use With (Application) Material Disposal
For Use With (Equipment) Binstands
Material Polyethylene
Sterility Non-sterile
Thickness (Metric) 0.08 mm
Size 202.28 x 165.1 cm
Thickness (English) 3 mL
Dimensions (L x W x H) 6 x 46 x 36 in.
Length (English) 46 in.
Length (Metric) 46 in.
Width (English) 36 in.
Width (Metric) 36 in.
Height (English) 6 in.
Height (Metric) 6 in.
Product Type Flat Pouch