missing translation for 'onlineSavingsMsg'
Learn More

BIN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20186-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889694

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

Involved in cytokinesis and septation where it has a role in the localization of F-actin.
Specifications

Specifications

BIN3
Polyclonal
Unconjugated
BIN3
BIN3, bridging integrator 3
Rabbit
Antigen Affinity Chromatography
RUO
361065, 55909, 57784
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q68FW8, Q9JI08, Q9NQY0
Bin3
BIN3 Fusion Protein Ag14091
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers