missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20186-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Involved in cytokinesis and septation where it has a role in the localization of F-actin.Specifications
BIN3 | |
Polyclonal | |
Unconjugated | |
BIN3 | |
BIN3, bridging integrator 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361065, 55909, 57784 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FW8, Q9JI08, Q9NQY0 | |
Bin3 | |
BIN3 Fusion Protein Ag14091 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |