Learn More
Emma Safety Footwear Billy XD Safety Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Specifications
| Design | Billy XD |
| Material | Leather |
| Size | 49 |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido