Learn More
Thermo Scientific™ Bile Esculin Azide Agar
Product Code. 10382223
Ansicht ändern
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Bile Esculin Azide Agar
Packungsgröße:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Format Media Type unitSize
10382223 10/Pk. 90mm Monoplate Bile Esculin Azide Agar Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10382223 Lieferant Thermo Scientific™ Lieferanten-Nr. PO5062A
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Isolate, enumerate and presumptively identify group D streptococci and enterococci with a medium that utilizes bile salts to improve selectivity.
Isolate, enumerate and presumptively identify group D streptococci and enterococci using Thermo Scientific™ Bile Esculin Azide Agar, a microbiological media that includes a high portion of Ox bile.
Ox bile present in the media inhibits the growth of Gram-positive bacteria except Enterococci, while Sodium azide suppresses the Gram-negative bacteria. Group D streptococci and enterococci are able to hydrolyse aesculin into aesculetin and glucose, and the bondage of aesculetin and ferric ammonium citrate causes the brown-black to black halo around the colonies.
Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Spezifikation
Color Honey yellow, transparent
Content And Storage 6°C to 12°C
Format 90mm Monoplate
Manufacturing Origin Germany
Media Type Bile Esculin Azide Agar
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.