Learn More
Invitrogen™ Bigfoot™ Spectral Cell Sorter Accessories
Product Code. 18074110
Change view
Click to view available options
Product Type:
Allergenco-D Cassette Holding Fixture
Autoclavable Cleaner Cap
Autoclavable Decon Cap
Autoclavable Filter, 0.05 μm
Bandpass Filter, 525/35
Chip Adapter, 10X
Full Decon NADCC Kit
Nozzle Tip, 100 μm
Nozzle Tip, 120 μm
Nozzle Tip, 150 μm
Nozzle Tip, 70 μm
Output Tube Adapter, 1.5 mL
Output Tube Adapter, 15 mL
Output Tube Adapter, 5 mL
Output Tube Adapter, 50 mL
Unit Size:
1 set
Each
Product Code. Product Type unitSize
18074110 Output Tube Adapter, 5 mL Each
18094110 Autoclavable Cleaner Cap Each
18014110 Autoclavable Decon Cap Each
17932051 Autoclavable Filter, 0.05 μm Each
18895490 Bandpass Filter, 525/35 Each
18054110 Chip Adapter, 10X Each
17414963 Nozzle Tip, 70 μm Each
17424963 Nozzle Tip, 100 μm Each
18851362 Nozzle Tip, 120 μm -
18841362 Nozzle Tip, 150 μm -
18064050 Output Tube Adapter, 1.5 mL Each
18074050 Output Tube Adapter, 15 mL Each
17464973 Output Tube Adapter, 50 mL Each
31722901 Full Decon NADCC Kit 1 set
31759049 Allergenco-D Cassette Holding Fixture Each
15 options
Product Code. 18074110
Brand: Invitrogen™ PL00356
The Bigfoot Cell Sorter has a multitude of cell sorting accessories to enable the most complex multi-parameter sorts and to meet high-throughput demands for a wide range of applications.
The Invitrogen Bigfoot Spectral Cell Sorter provides power, performance, and flexibility to help accelerate cutting edge discoveries. Designed to streamline the cell sorting workflow and enhance the user experience, the Bigfoot sorter has a multitude of cell sorting advancements, accessories, and unique capabilities to enable the most complex multi-parameter sorts and to meet high-throughput demands for a wide range of applications.
The Bigfoot Spectral Cell Sorter offers a full line of accessory products for continuous expansion of the instrument, including interchangeable nozzle tips, temperature controlled sample input and sort output adapters, and bulk fluid bottles and caps.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 mL
For Use With (Equipment) Bigfoot™ Spectral Cell Sorter
For Use With (Application) Flow Cytometry
Quantity Each
Content And Storage Room Temperature
Product Type Output Tube Adapter, 5 mL
Product Line Bigfoot
For Research Use Only.