missing translation for 'onlineSavingsMsg'
Learn More

Bicine, 99.38%, MP Biomedicals™

Catalog Number p-4403872
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
150-25-4
C6H12NNaO4
185.16
MFCD00004295
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
N, N-bis [2-hydroxyethyl]glycine
8761
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O
This item is not returnable. View return policy
150-25-4
C6H12NNaO4
185.16
MFCD00004295
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
N, N-bis [2-hydroxyethyl]glycine
8761
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 0.9938
IUPAC Name sodium 2-[bis(2-hydroxyethyl)amino]acetate
Formula Weight 163.2
Chemical Name or Material Bicine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'