missing translation for 'onlineSavingsMsg'
Learn More

BICD2 Antibody (1824), Novus Biologicals™

Product Code. 18100249 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18100249 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18100249 Supplier Novus Biologicals Supplier No. NBP243684

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunoprecipitation

BICD2 Monoclonal specifically detects BICD2 in Human, Rat samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BICD2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 1824
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunoprecipitation 1:100-1:500
Formulation PBS with No Preservative
Gene Alias bicaudal D homolog 2 (Drosophila), KIAA0699
Gene Symbols BICD2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human BICD2. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23299
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.