Learn More
Bibbette™ Pipette Adapter Support
Product Code. 12331228
Click to view available options
Length (Metric):
49mm
Unit Size:
Each
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12331228
Brand: Azlon PFL007
Only null left
Add to basket
Edge
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Specifications
Length (English) 1.92in.
Length (Metric) 49mm
Material Polypropylene
Product Type Bibbette™ pipette adapter support