Learn More
BIA 10-2474, MedChemExpress
Product Code. 16467878
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467878 10 mg 10mg
16443827 50 mg 50mg
16417696 100 mg 100mg
16400348 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16467878 Supplier Medchem Express Supplier No. HY19740
MedChemExpress BIA 10-2474 is an inhibitor of fatty acid amide hydrolase (FAAH) with IC50 values of 50 to 70mg/kg in various rat brain regions.
BIA 10-2474 is an inhibitor of fatty acid amide hydrolase (FAAH) with IC50 values of 50 to 70mg/kg in various rat brain regions.
Specifications
Chemical Name or Material BIA 10-2474
CAS 1233855-46-3
Color White
Purity Grade Notes Research
Molecular Formula C16H20N4O2
Quantity 10 mg
Solubility Information DMSO : 2.2 mg/mL (7.32 mM; Need ultrasonic)
SMILES O=C(N1C=C(C2=C[N+]([O-])=CC=C2)N=C1)N(C3CCCCC3)C
Molecular Weight (g/mol) 300.36
Formula Weight 300.36
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation