Learn More
Ansell™ Bi-Colour™ 87-900 Series Natural Rubber Latex Immersion Gloves
Product Code. 11731573 Shop All Ansell Products
Change view
Click to view available options
Size:
7.5
8
9
10
11
Unit Size:
12 pairs
Product Code. Size unitSize
11731573 7.5 12 pairs
11712833 8 12 pairs
11746883 9 12 pairs
11741573 10 12 pairs
11722833 11 12 pairs
5 options
Product Code. 11731573
Brand: Ansell™ 87900080
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With a latex/neoprene blend for reliable protection against a very wide range of chemicals. Ansell Bi-Colour™ 87-900 Series Natural Rubber Latex Immersion Gloves are double dipped for double protection.
  • Heavyweight resistance to salts and detergents
  • Quality flocklining in pure cotton to help reduce the risk of skin irritation
  • With embossed lozenge finish, green and yellow in color
  • Strong and durable, with excellent wet or dry grip
  • Ideal for aircraft assembly, battery and electronic manufacture, and the chemical industry
TRUSTED_SUSTAINABILITY
Specifications
Back Material Rubber Latex
Cuff Material Neoprene
Applications Industrial
Coating Material Natural Rubber Latex
Color Green
Latex Free No
Material Rubber Latex
Palm Thickness (English) 0.0268 in.
Palm Thickness (Metric) 0.68 mm
Product Type Immersion Gloves
Size 7.5
Thickness (English) 0.68 mm
Thickness (Metric) 0.68 mm
Glove Cuff Color Yellow
Glove Cuff Style Straight
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Salts and detergents resistant
Series 87-900