missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHMT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16351-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Homocysteine is a sulfur-containing amino acid that plays a crucial role in methylation reactions. Transfer of the methyl group from betaine to homocysteine creates methionine, which donates the methyl group to methylate DNA, proteins, lipids, and other intracellular metabolites. The protein encoded by this gene is one of two methyl transferases that can catalyze the transfer of the methyl group from betaine to homocysteine. Anomalies in homocysteine metabolism have been implicated in disorders ranging from vascular disease to neural tube birth defects such as spina bifida.Specifications
BHMT2 | |
Polyclonal | |
Unconjugated | |
BHMT2 | |
BHMT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23743, 365972, 64918 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q68FT5, Q91WS4, Q9H2M3 | |
Bhmt2 | |
BHMT2 Fusion Protein Ag9558 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |