missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHLHE40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522043
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
Specifications
| BHLHE40 | |
| Polyclonal | |
| Unconjugated | |
| BHLHE40 | |
| 1-Dec; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family member e40; basic helix-loop-helix family, member e40; bHLHb2; Bhlhe40; C130042M06Rik; Class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; Clast5; CR8; cytokine response gene 8; DEC1; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of split and hairy-related protein 2; enhancer-of-split and hairy-related protein 2; FLJ99214; HLHB2; SHARP2; SHARP-2; stimulated by retinoic acid 14; stimulated by retinoic acid gene 13 protein; STRA13; Stra14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O14503 | |
| BHLHE40 | |
| Synthetic peptide corresponding to a region within amino acids 1 and 81 of SHARP2 (Uniprot ID#O14503) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction