Learn More
Keystone Cleanroom Products BHD Blue Autoclaveable Wrapper
BIOPHARMAGREENER_CHOICE
Product Code. 17718197 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
8 x 8 in.
9 x 9 in.
12 x 12 in.
15 x 15 in.
18 x 18 in.
20 x 20 in.
24 x 24 in.
30 x 30 in.
36 x 36 in.
40 x 40 in.
45 x 45 in.
45 x 48 in.
54 x 54 in.
54 x 72 in.
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Product Code. 17718197
Brand: Keystone Cleanroom Products BHDSHEET3030
Only null left
Add to basket
Add to basket
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an adequate microbial barrier from external sources of contamination.
  • Designed for steam sterilization
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO Certified Cleanrooms
  • Certificate of Conformance included with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type None
Color Blue
Disposable Single-use
Material Cellulose
Sterility Non-sterile
Latex Free Yes
Dimensions (L x W) 30 x 30 in.
Length (English) 30 in.
Length (Metric) 76.2 cm
Width (English) 30 in.
Width (Metric) 76.2 cm
Product Type Sterilization Wrap (Heavy-duty)